missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protein 141 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane protein 141, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6803-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116317.1 | |
Transmembrane protein 141 Protein | |
Research Use Only | |
TMEM141 | |
Recombinant Protein | |
E. coli | |
MVNLGLSRVD DAVAAKHPGL GEYAACQSHA FMKGVFTFVT GTGMAFGLQM FIQRKFPYPL QWSLLVAVVA GSVVSYGVTR VESEKCNNLW LFLETGQLPK DRSTDQRS | |
Greater than 90% as determined by SDS-PAGE. |
85014 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |