missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protein 44 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane protein 44, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6806-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001011655.1 | |
Transmembrane protein 44 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
93109 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TMEM44 | |
Recombinant Protein | |
E. coli | |
MKSKMRQALG FAKEARESPD TQALLTCAEK EEENQENLDW VPLTTLSHCK SLRTMTAISR YMELTIEPVQ QAGCSATRLP GDGQTSAGDA SLQDPPSYPP VQVIRARVSS GSSSEVSSIN SDLEQKYWEA LNSEQWDPED VNLEGSKENV ELLGSQVHQD SVRTAHLSDD D |