missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRAPPC2 Protein
A cDNA sequence encoding the TRAPPC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13803-10ug
Additional Details : Weight : 0.01000kg
Specifications
6399 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGSFYF VIVGHHDNPV FEMEFLPAGK AESKDDHRHL NQFIAHAALD LVDENMWLSN NMYLKTVDKF NEWFVSAFVT AGHMRFIMLH DIRQEDGIKN FFTDVYDLYI KFSMNPFYEP NSPIRSSAFD RKVQFLGKKH LLS | |
Greater than 90% as determined by SDS-PAGE. |
TRAPPC2 Protein | |
Research Use Only | |
TRAPPC2 | |
Human | |
His | |
TRAPPC2 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 1mM DTT. |