missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRDMT1 Protein
A cDNA sequence encoding the TRDMT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13807-2ug
Additional Details : Weight : 0.01000kg
Specifications
TRDMT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEPLRV LELYSGVGGM HHALRESCIP AQVVAAIDVN TVANEVYKYN FPHTQLLAKT IEGITLEEFD RLSFDMILMS PPCQPFTRIG RQGDMTDSRT NSFLYILDIL PRLQKLPKYI LLENVKGFEV SSTRDLLIQT IENCGFQYQE FLLSPTSLGI PNSRLRYFLI AKLQSEPLPF QAPGQVLMEF PKIESVHPQK YAMDVENKIQ EKNVEPNISF DGSIQCSGKD AILFKLETAE EIHRKNQQDS DLSVKMLKDF LEDDTDVNQY LLPPKSLLRY ALLLDIVQPT CRRSVCFTKG YGSYIEGTGS VLQTAEDVQV ENIYKSLTNL SQEEQITKLL ILKLRYFTPK EIANLLGFPP EFGFPEKITV KQRYRLLGNS LNVHVVAKLI KILYE | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TRDMT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |