missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TREM2 Protein

A DNA sequence encoding the Homo sapiens (Human) TREM2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6839-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036200

  • 505.00 € / 100µg
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001258750.1
TREM2 Protein
Research Use Only
Trem2
Recombinant Protein
E. coli
HNTTVFQGVA GQSLQVSCPY DSMKHWGRRK AWCRQLGEKG PCQRVVSTHN LWLLSFLRRW NGSTAITDDT LGGTLTITLR NLQPHDAGLY QCQSLHGSEA DTLRKVLVEV LADPLDHRDA GDLWFPGESE SFEDAHVEHS ISRSLLEGEI PFPPTS
Greater than 90% as determined by SDS-PAGE.
54209
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers