missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRIP10 Protein
A cDNA sequence encoding the TRIP10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13817-20ug
Additional Details : Weight : 0.01000kg
Specifications
TRIP10 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TRIP10 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol, 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDPKNDSH VLIELHKSGF ARPGDVEFED FSQPMNRAPS DSSLGTPSDG RPELRGPGRS RTKRWPFGKK NKTVVTEDFS HLPPEQQRKR LQQQLEERSR ELQKEVDQRE ALKKMKDVYE KTPQMGDPAS LEPQIAETLS NIERLKLEVQ KYEAWLAEAE SRVLSNRGDS LSRHARPPDP PASAPPDSSS NSASQDTKES SEEPPSEESQ DTPIYTEFDE DFEEEPTSPI GHCVAIYHFE GSSEGTISMA EGEDLSLMEE DKGDGWTRVR RKEGGEGYVP TSYLRVTLN |