missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tryptase beta-2 Protein

Product Code. 16016150
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. Quantity unitSize
16016150 1 mg 1mg
16006150 10 μg 10µg
16046150 50 μg 50µg
16096140 100 μg 100µg
16026150 200 μg 200µg
16036150 500 μg 500µg
6 options
This item is not returnable. View return policy

Product Code. 16016150

Brand: enQuireBio™ QP6832ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Tryptase beta-2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_003285.2
Gene ID (Entrez) 64499
Name Tryptase beta-2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TPSB2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence IVGGQEAPRS KWPWQVSLRV HGPYWMHFCG GSLIHPQWVL TAAHCVGPDV KDLAALRVQL REQHLYYQDQ LLPVSRIIVH PQFYTAQIGA DIALLELEEP VNVSSHVHTV TLPPASETFP PGMPCWVTGW GDVDNDERLP PPFPLKQVKV PIMENHICDA KYHLGAYTGD DVRIVRDDML CAGNTRRDSC QGDSGGPLVC KVNGTWLQAG VVSWGEGCAQ PNRPGIYTRV TYYLDWIHHY VPKKP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.