missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tubulin polymerization-promoting Protein

A DNA sequence encoding the Homo sapiens (Human) Tubulin polymerization-promoting protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6830-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036130

  • 2365.00 € / 1mg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_008961.1
Tubulin polymerization-promoting Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
11076
Greater than 80% as determined by SDS-PAGE.
Research Use Only
TPPP
Recombinant Protein
E. coli
MADKAKPAKA ANRTPPKSPG DPSKDRAAKR LSLESEGAGE GAAASPELSA LEEAFRRFAV HGDARATGRE MHGKNWSKLC KDCQVIDGRN VTVTDVDIVF SKIKGKSCRT ITFEQFQEAL EELAKKRFKD KSSEEAVREV HRLIEGKAPI ISGVTKAISS PTVSRLTDTT KFTGSHKERF DPSGKGKGKA GRVDLVDESG YVSGYKHAGT YDQKVQGGK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers