missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tumor protein D52 Protein

A DNA sequence encoding the Homo sapiens (Human) Tumor protein D52, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6825-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036100

  • 2365.00 € / 1mg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001020423.1
Tumor protein D52 Protein
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
7163
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TPD52
Recombinant Protein
E. coli
MDRGEQGLLR TDPVPEEGED VAATISATET LSEEEQEELR RELAKVEEEI QTLSQVLAAK EKHLAEIKRK LGINSLQELK QNIAKGWQDV TATSAYKKTS ETLSQAGQKA SAAFSSVGSV ITKKLEDVKN SPTFKSFEEK VENLKSKVGG TKPAGGDFGE VLNSAANASA TTTEPLPEKT QESL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers