missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Twinkle protein, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Twinkle protein, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9861-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048871

  • 1665.00 € / 500µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001157284.1
Twinkle protein, mitochondrial Protein
Research Use Only
PEO1
Recombinant Protein
Yeast
APGPPRRRYR KETLQALDMP VLPVTATEIR QYLRGHGIPF QDGHSCLRAL SPFAESSQLK GQTGVTTSFS LFIDKTTGHF LCMTSLAEGS WEDFQASVEG RGDGAREGFL LSKAPEFEDS EEVRRIWNRA IPLWELPDQE EVQLADTMFG LTKVTDDTLK RFSVRYLRPA RSLVFPWFSP GGSGLRGLKL LEAKCQGDGV SYEETTIPRP SAYHNLFGLP LISRRDAEVV LTSRELDSLA LNQSTGLPTL TLPRGTTCLP PALLPYLEQF RRIVFWLGDD LRSWEAAKLF ARKLNPKRCF LVRPGDQQPR PLEALNGGFN LSRILRTALP AWHKSIVSFR QLREEVLGEL SNVEQAAGLR WSRFPDLNRI LKGHRKGELT VFTGPTGSGK TTFISEYALD LCSQGVNTLW GSFEISNVRL ARVMLTQFAE GRLEDQLDKY DHWADRFEDL PLYFMTFHGQ QSIRTVIDTM QHAVYVYDIC HVIIDNLQFM MGHEQLSTDR IAAQDYIIGV FRKFATDNNC HVTLVIHPRK EDDDKELQTA SIFGSAKASQ EADNVLILQD RKLVTGPGKR YLQVSKNRFD GDVGVFPLEF NKNSLTFSIP PKNKARLKKI KDDTGPVAKK PSSGKKGATT QNSEICSGQA PTPDQPDTSK RSK
Greater than 90% as determined by SDS-PAGE.
56652
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers