missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TYW5 Protein
A cDNA sequence encoding the TYW5 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | TYW5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994889
|
enQuireBio™
QP13847-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984889
|
enQuireBio™
QP13847-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974889
|
enQuireBio™
QP13847-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TYW5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TYW5 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGQHLP VPRLEGVSRE QFMQHLYPQR KPLVLEGIDL GPCTSKWTVD YLSQVGGKKE VKIHVAAVAQ MDFISKNFVY RTLPFDQLVQ RAAEEKHKEF FVSEDEKYYL RSLGEDPRKD VADIRKQFPL LKGDIKFPEF FKEEQFFSSV FRISSPGLQL WTHYDVMDNL LIQVTGKKRV VLFSPRDAQY LYLKGTKSEV LNIDNPDLAK YPLFSKARRY ECSLEAGDVL FIPALWFHNV ISEEFGVGVN IFWKHLPSEC YDKTDTYGNK DPTAASRAAQ ILDRALKTLA ELPEEYRDFY ARRMVLHIQD KAYSKNSE | |
Greater than 95.0% as determined by SDS-PAGE. |