missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human U2 small nuclear ribonucleoprotein B'' Protein

A DNA sequence encoding the Homo sapiens (Human) U2 small nuclear ribonucleoprotein B'', was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7374-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048540

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003083.1
U2 small nuclear ribonucleoprotein B'' Protein
Research Use Only
SNRPB2
Recombinant Protein
E. coli
MDIRPNHTIY INNMNDKIKK EELKRSLYAL FSQFGHVVDI VALKTMKMRG QAFVIFKELG SSTNALRQLQ GFPFYGKPMR IQYAKTDSDI ISKMRGTFAD KEKKKEKKKA KTVEQTATTT NKKPGQGTPN SANTQGNSTP NPQVPDYPPN YILFLNNLPE ETNEMMLSML FNQFPGFKEV RLVPGRHDIA FVEFENDGQA GAARDALQGF KITPSHAMKI TYAKK
Greater than 90% as determined by SDS-PAGE.
6629
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers