missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2D2 Protein
A cDNA sequence encoding the UBE2D2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | UBE2D2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984919
|
enQuireBio™
QP13856-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974919
|
enQuireBio™
QP13856-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964919
|
enQuireBio™
QP13856-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
UBE2D2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The UBE2D2 solution (0.5 mg/ml) contains 20mM MES (pH6.0), 50mM NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MALKRIHKEL NDLARDPPAQ CSAGPVGDDM FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TRIYHPNINS NGSICLDILR SQWSPALTIS KVLLSICSLL CDPNPDDPLV PEIARIYKTD REKYNRIARE WTQKYAM | |
Greater than 95% as determined by SDS-PAGE. |