missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2T Protein

A cDNA sequence encoding the UBE2T was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13868-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934959

  • 203.00 € / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UBE2T Protein
Research Use Only
Recombinant Protein
E. coli
MQRASRLKRE LHMLATEPPP GITCWQDKDQ MDDLRAQILG GANTPYEKGV FKLEVIIPER YPFEPPQIRF LTPIYHPNID SAGRICLDVL KLPPKGAWRP SLNIATVLTS IQLLMSEPNP DDPLMADISS EFKYNKPAFL KNARQWTEKH ARQKQKADEE EMLDNLPEAG DSRVHNSTQK RKASQLVGIE KKFHPDVLEH HHHHH
Greater than 95% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The UBE2T solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers