missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UFC1 Protein
A cDNA sequence encoding the UFC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13881-5ug
Additional Details : Weight : 0.01000kg
Specifications
UFC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADEATRRVV SEIPVLKTNA GPRDRELWVQ RLKEEYQSLI RYVENNKNAD NDWFRLESNK EGTRWFGKCW YIHDLLKYEF DIEFDIPITY PTTAPEIAVP ELDGKTAKMY RGGKICLTDH FKPLWARNVP KFGLAHLMAL GLGPWLAVEI PDLIQKGVIQ HKEKCNQ | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UFC1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |