missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UFD1L, homolog Protein

A DNA sequence encoding the Homo sapiens (Human) UFD1L, homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7910-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031541

  • 2285.00 € / 1mg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001030324.2
UFD1L, homolog Protein
Research Use Only
UFD1L
Recombinant Protein
E. coli
MFSFNMFDHP IPRVFQNRFS TQYRCFSVSM LAGPNDRSDV EKGGKIIMPP SALDQLSRLN ITYPMLFKLT NKNSDRMTHC GVLEFVADEG ICYLPHWMMQ NLLLEEGGLV QVESVNLQVA TYSKFQPQSP DFLDITNPKA VLENALRNFA CLTTGDVIAI NYNEKIYELR VMETKPDKAV SIIECDMNVD FDAPLGYKEP ERQVQHEEST EGEADHSGYA GELGFRAFSG SGNRLDGKKK GVEPSPSPIK PGDIKRGIPN YEFKLGKITF IRNSRPLVKK VEEDEAGGRF VAFSGEGQSL RKKGRKP
Greater than 80% as determined by SDS-PAGE.
7353
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers