missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Uncharacterized protein C20orf197 Protein

A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C20orf197, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7850-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041171

  • 2285.00 € / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001289742.1
Uncharacterized protein C20orf197 Protein
Research Use Only
C20orf197
Recombinant Protein
E. coli
MVALFQHSPY WQADGYGHSH RLKCQHFKQH RQYNDKLEIS SNLGPQFNAL LNILLNIVHP TLSHDTRRSK GLKIEGLLQS RELGNSWTVT MCIWVLKALQ SSAPNKPLDW LDPMPCFQNL LARGTP
Greater than 90% as determined by SDS-PAGE.
284756
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers