missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C4orf3 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C4orf3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7860-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001701.2 | |
Uncharacterized protein C4orf3 Protein | |
Research Use Only | |
C4orf3 | |
Recombinant Protein | |
E. coli | |
MEVDAPGVDG RDGLRERRGF SEGGRQNFDV RPQSGANGLP KHSY | |
Greater than 90% as determined by SDS-PAGE. |
401152 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |