missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Urokinase / PLAU Protein
A DNA sequence encoding the Homo sapiens (Human) Urokinase / PLAU, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8792-ec-1mg
Informations supplémentaires : Weight : 0.01000kg
Spécification
NP_001138503.1 | |
Urokinase / PLAU Protein | |
Research Use Only | |
PLAU | |
Recombinant Protein | |
E. coli | |
SNELHQVPSN CDCLNGGTCV SNKYFSNIHW CNCPKKFGGQ HCEIDKSKTC YEGNGHFYRG KASTDTMGRP CLPWNSATVL QQTYHAHRSD ALQLGLGKHN YCRNPDNRRR PWCYVQVGLK PLVQECMVHD CADGKKPSSP PEELKFQCGQ KTL | |
Greater than 90% as determined by SDS-PAGE. |
5328 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |