missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAMP4 Protein
A cDNA sequence encoding the VAMP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13918-10ug
Additional Details : Weight : 0.01000kg
Specifications
VAMP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPPKFKRHLN DDDVTGSVKS ERRNLLEDDS DEEEDFFLRG PSGPRFGPRN DKIKHVQNQV DEVIDVMQEN ITKVIERGER LDELQDKSES LSDNATAFSN RSKQLRRQMW WRGCKLEHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAMP4 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, and 20% glycerol. |