missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VDR Protein
A cDNA sequence encoding the VDR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13932-20ug
Additional Details : Weight : 0.01000kg
Specifications
VDR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSYYHHHHHHDYDIPTTDYKDDDDKDYKDDDDKENLYFQGEFMEAMAASTSLPDPGDFDRNVPRICGVCGDRATGFHFNAMTCEGCKGFFRRSMKRKALFTCPFNGDCRITKDNRRHCQACRLKRCVDIGMMKEFILTDEEVQRKREMILKRKEEEALKDSLRPKLSEEQQRIIAILLDAHHKTYDPTYSDFCQFRPPVRVNDGGGSHPSRPNSRHTPSFSGDSSSSCSDHCITSSDMMDSSSFSNLDLSEEDSDDPSVTLELSQLSMLPHLADLVSYSIQKVIGFAKMIPGFRDLTSEDQIVLLKSSAIEVIMLRSNESFTMDDMSWTCGNQDYKYRVSDVTKAGHSLELIEPLIKFQVGLKKLNLHEEEHVLLMAICIVSPDRPGVQDAALIEAIQDRLSNTLQTYIRCRHPPPGSHLLYAKMIQKLADLRSLNEEHSKQYRCLSFQPECSMKLTPLVLEVFGNEIS | |
Greater than 70.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Flag Tag | |
VDR protein is supplied in 50mM Tris, 150mM NaCl and 10% Glycerol, pH 7.5. |