missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VWA5B2 Protein

A DNA sequence encoding the Homo sapiens (Human) VWA5B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7823-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051011

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001307302.1
VWA5B2 Protein
Research Use Only
VWA5B2
Recombinant Protein
E. coli
PRKPSLGAIL DGPSPEPGQQ LGQGLDDSGN LLSPAPMDWD MLMEPPFLFT AVPPSGELAP PAVPPQAPRC HVVIRGLCGE QP
Greater than 90% as determined by SDS-PAGE.
90113
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers