missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WIF1 Protein
A cDNA sequence encoding the WIF1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13959-5ug
Additional Details : Weight : 0.01000kg
Specifications
11197 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADLGPPQEES LYLWIDAHQA RVLIGFEEDI LIVSEGKMAP FTHDFRKAQQ RMPAIPVNIH SMNFTWQAAG QAEYFYEFLS LRSLDKGIMA DPTVNVPLLG TVPHKASVVQ VGFPCLGKQD GVAAFEVDVI VMNSEGNTIL KTPQNAIFFK TCQQAECPGG CRNGGFCNER RICECPDGFH GPHCEKALCT PRCMNGGLCV TPGFCICPPG FYGVNCDKAN CSTTCFNGGT CFYPGKCICP PGLEGEQCEI SKCPQPCRNG GKCIGKSKCK CSKGYQGDLC SKPVCEPGCG AHGTCHEPNK CQCQEGWHGR HCNKRYEASL IHALRPAGAQ LRQHTPSLKK AEERRDPPES NYIWSGRLVP RGSHHHHHH | |
Greater than 80.0% as determined by SDS-PAGE. |
WIF1 Protein | |
Research Use Only | |
WIF1 | |
Human | |
His | |
The WIF1 protein solution contains 1X PBS pH 7.4, 10% glycerol and 1mM PMSF. |