missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YWHAG Protein
A cDNA sequence encoding the YWHAG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13986-HIS-2ug
Additional Details : Weight : 0.01000kg
Specifications
7532 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
YWHAG | |
Human | |
His | |
YWHAG solution containing 20mM Tris 7.5. |
YWHAG Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MVDREQLVQK ARLAEQAERY DDMAAAMKNV TELNEPLSNE ERNLLSVAYK NVVGARRSSW RVISSIEQKT SADGNEKKIE MVRAYREKIE KELEAVCQDV LSLLDNYLIK NCSETQYESK VFYLKMKGDY YRYLAEVATG EKRATVVESS EKAYSEAHEI SKEHMQPTHP IRLGLALNYS VFYYEIQNAP EQACHLAKTA FDDAIAELDT LNEDSYKDST LIMQLLRDNL TLWTSDQQDD DGGEGNN |