missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein
A DNA sequence encoding the Influenza A virus (strain A/Fort Warren/1/1950 H1N1) Influenza A H1N1 (strain A/Fort Warren/1/1950) Nucleocapsid, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Name | Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NP |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047320
|
enQuireBio™
QP7081-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087320
|
enQuireBio™
QP7081-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037320
|
enQuireBio™
QP7081-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067320
|
enQuireBio™
QP7081-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077320
|
enQuireBio™
QP7081-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057320
|
enQuireBio™
QP7081-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza A H1N1 (Fort Warren / 1 / 1950) Nucleocapsid Protein | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGDRQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVSSFIRGT KVIPRGKLST RGVQIASNEN MDTMGSSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DKTTIMAAFT GNAEGRTSDM RAEIIRMMES ARPEEVSFQG RGVFELSDER AANPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |