Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Wilson-Smith / 1933) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Wilson-Smith/1933 H1N1) Influenza A H1N1 (strain A/Wilson-Smith/1933) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7106-100ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H1N1 (Wilson-Smith / 1933) NucleoProtein | |
100 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MATKGTKRSY EQMETDGERQ NATEIRASVG KMIGGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYRRV DGKWMRELIL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRRTRIAYE RMCNILKGKF QTAAQRAMVD QVRESRNPGN AEFEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEREGYSLVG IDPFRLLQNS QVYSLIRPNE NPAHKSQLVW MACHSAAFED LRVSSFIRGT KVVPRGKLST RGVQIASNEN METMESSTLE LRSRYWAIRT RSGGNTNQQR ASSGQISIQP TFSVQRNLPF DRPTIMAAFT GNTEGRTSDM RTEIIRLMES ARPEDVSFQG RGVFELSDEK AASPIVPSFD MSNEGSYFFG DNAEEYDN |