missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A Influenza A HA Protein
Recombinant Protein
385.00 € - 2210.00 €
Specifications
Name | Influenza A Influenza A HA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964008
|
enQuireBio™
QP10486-100UG |
100 μg |
385.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984008
|
enQuireBio™
QP10486-500UG |
500 μg |
1355.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974008
|
enQuireBio™
QP10486-1MG |
1 mg |
2210.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza A Influenza A HA Protein | |
Untested | |
Influenza A | |
His | |
>95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
PBS pH7.4. |