missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Influenza A Virus H3N2 Hemagglutinin Protein

Product Code. 18796976
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796976 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796976 Supplier R&D Systems™ Supplier No. 10944HA100
  • ONLINE EXCLUSIVE PRICE
    546.34 €
    487.20 € / 100µg
    Save 59.14 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

His-tag

Measured by its binding ability in a functional ELISA. When Recombinant Human Galectin-1 (Catalog # 1152-GA ) immobilized at 10.00 μg/mL, 100 μL/well, the concentration of Recombinant Influenza A Virus H3N2 Hemagglutinin His-tag (Catalog # 10944-HA) that produces 50% of the optimal binding response is 0.35-3.50 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q91MA7.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) MolecularWeight-observed: 80-90 kDa, under reducing conditions., MolecularWeight-theroretical: 58 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived influenza a virus h3n2 Hemagglutinin protein Gln17-Trp530, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Hemagglutinin/HA Peptide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.