missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Jilin Protein
A cDNA sequence encoding the Influenza-B Jilin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12439-100ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza-B Jilin Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
The Recombinant B/Jilin/20/2003 solution contains 10mM Sodium phosphate, pH 7.4 and 150mM Sodium Cloride. |