missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza-B Malaysia Protein

A cDNA sequence encoding the Influenza-B Malaysia was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12440-INSECT-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930429

  • 288.00 € / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza-B Malaysia Protein
Research Use Only
Recombinant Protein
Baculovirus
The Recombinant B/Malaysia/2506/2004 solution contains 10mM Sodium Phosphate, pH 7.0 and 150mM NaCl, 0.005% Tween-20.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers