missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza-B Victoria Protein

A cDNA sequence encoding the Influenza-B Victoria was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12444-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940439

  • 131.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza-B Victoria Protein
Research Use Only
Recombinant Protein
E. coli
The B/ Victoria /53/99 solution contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers