missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Victoria Protein
A cDNA sequence encoding the Influenza-B Victoria was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12444-1mg
Additional Details : Weight : 0.01000kg
Specifications
Influenza-B Victoria Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The B/ Victoria /53/99 solution contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |