missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza Florida 04/2006 Recombinant Protein

A cDNA sequence encoding the Florida 04/2006 Recombinant was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11889-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978698

  • 2610.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza Florida 04/2006 Protein
Research Use Only
Recombinant Protein
Baculovirus
The Recombinant B/ Florida 04/2006solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers