missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H1N1 New Caledonia Protein

A cDNA sequence encoding the H1N1 New Caledonia was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12076-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959288

  • 2610.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza H1N1 New Caledonia Protein
Research Use Only
Recombinant Protein
Influenza H1N1
The Recombinant H1N1 A/New Caledonia/20/99 solution 10mM Sodium phosphate, pH 7.2, 150mM NaCl.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by: Analysis by SDS-PAGE, HA1 and HA2 bands are observed using SDS-PAGE under reducing conditions.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers