missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 New Caledonia Protein
A cDNA sequence encoding the H1N1 New Caledonia was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12076-2ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza H1N1 New Caledonia Protein | |
Research Use Only | |
Recombinant Protein | |
Influenza H1N1 | |
The Recombinant H1N1 A/New Caledonia/20/99 solution 10mM Sodium phosphate, pH 7.2, 150mM NaCl. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by: Analysis by SDS-PAGE, HA1 and HA2 bands are observed using SDS-PAGE under reducing conditions. |