missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Protein
A cDNA sequence encoding the H3N2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | Influenza H3N2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939308
|
enQuireBio™
QP12080-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929308
|
enQuireBio™
QP12080-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919308
|
enQuireBio™
QP12080-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
His | |
The H3N2 Canine solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADNLPGNENN AATLCLGHHA VPNGTIVKTI TDDQIEVTNA TELVQNSSTG KICNNPHKIL DGRDCTLIDA LLGDPHCDVF QNETWDLFVE RSNAFSNCYP YDVPDYASLR SIVASSGTLE FITEGFTWAG VTQNGGSGAC KKGPANGFFS RLNWLTKSGN TYPVLNVTMP NNNNFDKLYI WGVHHPSTNQ EQTSLYIQAS GRVKVSTRRS QQTIIPNIGS RPLVRGQSGR ISVYWTIVKP GDVLVINSNG NLIAPRGYFK MRIGKSSIMR SDAPIDTCIS ECITPNGSIP NEKPFQNVNK ITYGACPKYV KQNTLKLATG MRNVPERQTH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |