missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Shandong Protein

Product Code. 15999318
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999318 10 μg 10µg
15919328 50 μg 50µg
15909328 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15999318 Supplier enQuireBio™ Supplier No. QP1208610ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Name Influenza H3N2 Shandong Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Influenza
Species Baculovirus
Protein Tag Untagged
Buffer The H3N2A/ Shandong/9/93 solution (1.0 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
Purity or Quality Grade Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.