missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Uruguay Recombinant Protein
A cDNA sequence encoding the H3N2 Uruguay Recombinant was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | Influenza H3N2 Uruguay Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909338
|
enQuireBio™
QP12089-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999328
|
enQuireBio™
QP12089-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989328
|
enQuireBio™
QP12089-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Uruguay Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H3N2 A/ Uruguay 716/2007solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20. |