missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H5N1 Vietnam Protein
A cDNA sequence encoding the H5N1 Vietnam was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | Influenza H5N1 Vietnam Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979348
|
enQuireBio™
QP12094-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969348
|
enQuireBio™
QP12094-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959348
|
enQuireBio™
QP12094-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H5N1 Vietnam Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
The Recombinant H5N1 A/Vietnam/1203/04 solution contains 10mM Sodium Phosphate, pH 7.1, 0.005% Tween-20 and 150mM NaCl. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
V K M E K I V L L F A I V S L V K S D Q I C I G Y H A N N S T E Q V D T I M EK N V T V T H A Q D I L E K K H N G K L C D L D G V K P L I L R D C S V A GW L L G N P M C D E F I N V P E W S Y I V E K A N P V N D L C Y P G D F N DY E E L K H L L S R I N H F E K I Q I I P K S S W S S H E A S L G V S S A C PY Q G K S S F F R N V V W L I K K N S T Y P T I K R S Y N N T N Q E D L L VL W G I H H P N D A A E Q T K L Y Q N P T T Y I S V G T S T L N Q R L V P R IA T R S K V N G Q S G R M E F F W T I L K P N D A I N F E S N G N F I A P EY A Y K I V K K G D S T I M K S E L E Y G N C N T K C Q T P M G A I N S S MP F H N I H P L T I G E C P K Y V K S N R L V L A T G L R N S P Q R E R R RK K R G L F G A I A G F I E G G W Q G M V D G W Y G Y H H S N E Q G S GY A A D K E S T Q K A I D G V T N K V N S I I D K M N T Q F E A V G R E F NN L E R R I E N L N K K M E D G F L D V W T Y N A E L L V L M E N E R T L DF H D S N V K N L Y D K V R L Q L R D N A K E L G N G C F E F Y H K C D NE C M E S V R N G T Y D Y | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE, HA1 and HA2 bands are observed using SDS-PAGE under reducing conditions. |