missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza New York 3571/2009 Protein
A cDNA sequence encoding the New York 3571/2009 was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | Influenza New York 3571/2009 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991689
|
enQuireBio™
QP12833-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981689
|
enQuireBio™
QP12833-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971689
|
enQuireBio™
QP12833-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza New York 3571/2009 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
His | |
The H1N1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLMDTLCIG YHANNSTDTV DTVLEKNVTV THSVNLLEDK HNGKLCKLRG VAPLHLGKCN IAGWILGNPE CESLSTASSW SYIVETSSSD NGTCYPGDFI DYEELREQLS SVSSFERFEI FPKTSSWPNH DSNKGVTAAC PHAGAKSFYK NLIWLVKKGN SYPKLSKSYI NDKGKEVLVL WGIHHPSTSA DQQSLYQNAD AYVFVGSSRY SKKFKPEIAI RPKVRDQEGR MNYYWTLVEP GDKITFEATG NLVVPRYAFA MERNAGSGII ISDTPVHDCN TTCQTPKGAI NTSLPFQNIH PITIGKCPKY VKSTKLRLAT GLRNVPSIQS RSRHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |