missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Vietnam HN31242/2007 Protein
A cDNA sequence encoding the Vietnam HN31242/2007 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13935-100ug
This item is not returnable.
View return policy
Specifications
Influenza Vietnam HN31242/2007 Protein | |
Research Use Only | |
H | |
Influenza | |
His | |
The H5N1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPMDQICIG YHANNSTEQV DTIMEKNVTV THAQDILEKT HNGKLCDLDG VKPLILRDCS VAGWLLGNPM CDEFINVPEW SYIVEKANPA NDLCYPGNFN DYEELKHLLS RINHFEKIQI IPKSSWSDHE ASSGVSSACP YQGVPSFFRN VVWLIKKNNT YPTIKRSYNN TNQEDLLILW GIHHSNDAAE QTKLYQNPTT YISVGTSTLN QRLVPKIATR SKVNGQSGRM DFFWTILKPN DAINFESNGN FIAPEYAYKI VKKGDSAIMK SEVEYGNCNT KCQTPIGAIN SSMPFHNIHP LTIGECPKYV KSNKLVLATG LRNSPLHHHH HH | |
Greater than 90% as determined by SDS-PAGE. |