missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL-1 alpha / IL1A / IL1F1 Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6213-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031620

  • 940.00 € / 100µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001036222.1
IL-1 alpha / IL1A / IL1F1 Protein
Research Use Only
IL1A
Recombinant Protein
E. coli
SAPFSFLSNM TYHFIRIIKH EFILNDTLNQ TIIRANDQHL TAAAIHNLDE AVKFDMGAYT SSKDDTKVPV ILRISKTQLY VSAQDEDQPV LLKEMPEINK TITGSETNFL FFWETHGTKN YFISVAHPNL FIATKHDNWV CLAKGLPSIT DFQILENQA
Greater than 90% as determined by SDS-PAGE.
700193
100 μg
Not determined.
Full Length
Macaque
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers