missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL-15-S48AA Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-15-S48AA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9208-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077851

  • 2535.00 € / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001038196.1
IL-15-S48AA Protein
Research Use Only
IL15
Recombinant Protein
Yeast
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SHESGDTDIH DTVENLIILA NNILSSNGNI TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS
Greater than 90% as determined by SDS-PAGE.
699616
500 μg
Not determined.
Full Length
Macaque
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers