missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL17 / IL17A Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL17 / IL17A, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | XP_001106391.1 |
---|---|
Gene ID (Entrez) | 708123 |
Name | IL17 / IL17A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051600
|
enQuireBio™
QP6211-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071600
|
enQuireBio™
QP6211-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061600
|
enQuireBio™
QP6211-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081600
|
enQuireBio™
QP6211-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091600
|
enQuireBio™
QP6211-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001610
|
enQuireBio™
QP6211-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
XP_001106391.1 | |
IL17 / IL17A Protein | |
Not determined. | |
Full Length | |
Macaque | |
GST | |
Tris-based buffer, 50% glycerol |
708123 | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
GIAIPRNPGC PNSEDKTFPR TVMVNLNIHN RNTNTNPKRS SDYYNRSTSP WNLHRNEDPE RYPSVIWEAK CRHLGCVNAD GNVDYHMNSV PIQQEILVLR REPRHCPNSF RLEKILVSVG CTCVTPIVHH VA | |
Greater than 90% as determined by SDS-PAGE. |