missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Acetylcholine receptor subunit alpha Protein

A DNA sequence encoding the Mus musculus (Mouse) Acetylcholine receptor subunit alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5833-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958099

  • 735.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_031415.2
Acetylcholine receptor subunit alpha Protein
Research Use Only
Chrna1
Recombinant Protein
E. coli
SEHETRLVAK LFEDYSSVVR PVEDHREIVQ VTVGLQLIQL INVDEVNQIV TTNVRLKQQW VDYNLKWNPD DYGGVKKIHI PSEKIWRPDV VLYNNADGDF AIVKFTKVLL DYTGHITWTP PAIFKSYCEI IVTHFPFDEQ NCSMKLGTWT YDGSVVAINP ESDQPDLSNF MESGEWVIKE ARGWKHWVFY SCCPTTPYLD ITYHFVMQRL
Greater than 80% as determined by SDS-PAGE.
11435
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers