missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse aFGF / FGF1 Protein
Recombinant Protein
107.00 € - 1885.00 €
Specifications
Accession Number | NP_034327.1 |
---|---|
Gene ID (Entrez) | 14164 |
Name | aFGF / FGF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976159
|
enQuireBio™
QP5455-10UG |
10 μg |
107.00 €
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906169
|
enQuireBio™
QP5455-50UG |
50 μg |
252.00 €
50µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966159
|
enQuireBio™
QP5455-100UG |
100 μg |
433.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996159
|
enQuireBio™
QP5455-500UG |
500 μg |
1110.00 €
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986159
|
enQuireBio™
QP5455-1MG |
1 mg |
1885.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034327.1 | |
aFGF / FGF1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse aFGF / FGF1 induces 3T3 cell proliferation (typical ED50 is around 0.1 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5 |
14164 | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE. |