missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse aFGF / FGF1 Protein
A DNA sequence encoding the Mus musculus (Mouse) aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6037-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034327.1 | |
aFGF / FGF1 Protein | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
FNLPLGNYKK PKLLYCSNGG HFLRILPDGT VDGTRDRSDQ HIQLQLSAES AGEVYIKGTE TGQYLAMDTE GLLYGSQTPN EECLFLERLE ENHYNTYTSK KHAEKNWFVG LKKNGSCKRG PRTHYGQKAI LFLPLPVSSD | |
Greater than 90% as determined by SDS-PAGE. |
14164 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |