missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse aFGF / FGF1 Protein
A DNA sequence encoding the Mus musculus (Mouse) aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_034327.1 |
---|---|
Gene ID (Entrez) | 14164 |
Name | aFGF / FGF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939529
|
enQuireBio™
QP6037-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959529
|
enQuireBio™
QP6037-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949529
|
enQuireBio™
QP6037-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969529
|
enQuireBio™
QP6037-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979529
|
enQuireBio™
QP6037-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989529
|
enQuireBio™
QP6037-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034327.1 | |
aFGF / FGF1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
14164 | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
FNLPLGNYKK PKLLYCSNGG HFLRILPDGT VDGTRDRSDQ HIQLQLSAES AGEVYIKGTE TGQYLAMDTE GLLYGSQTPN EECLFLERLE ENHYNTYTSK KHAEKNWFVG LKKNGSCKRG PRTHYGQKAI LFLPLPVSSD | |
Greater than 90% as determined by SDS-PAGE. |