missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Allograft inflammatory factor 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5631-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956569

  • 735.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062340.1
Allograft inflammatory factor 1 Protein
Research Use Only
Aif1
Recombinant Protein
E. coli
SQSRDLQGGK AFGLLKAQQE ERLEGINKQF LDDPKYSNDE DLPSKLEAFK VKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKRLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH KRPTGPPAKK AISELP
Greater than 90% as determined by SDS-PAGE.
11629
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers