missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Apolipoprotein 1

A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5668-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966859

  • 735.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001292748.1
Apolipoprotein 1
Research Use Only
Apoe
Recombinant Protein
E. coli
EGEPEVTDQL EWQSNQPWEQ ALNRFWDYLR WVQTLSDQVQ EELQSSQVTQ ELTALMEDTM TEVKAYKKEL EEQLGPVAEE TRARLGKEVQ AAQARLGADM EDLRNRLGQY RNEVHTMLGQ STEEIRARLS THLRKMRKRL MRDAEDLQKR LAVYKAGARE GAERGVSAIR ERLGPLVEQG RQRTANLGAG AAQPLRDRAQ AFGDRIRGRL EEVGNQARDR LEEVREHMEE VRSKMEEQTQ QIRLQAEIFQ ARLKGWFEPI VEDMHRQWAN LMEKIQASVA TNPIITPVAQ ENQ
Greater than 89.4% as determined by SDS-PAGE.
11816
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers